Oxazol-5-methanol
Lieferant: COMBI-BLOCKS INC.
COBBHI-1270-1GEA
369
EUR
COBBHI-1270-1G
COBBHI-1270-5G
Oxazol-5-methanol
Oxazol-5-methanol
Formel:
C₄H₅NO₂ Molecular Weight: 99,0880 g/mol |
MDL:
MFCD09027272 CAS-Nummer: 127232-41-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...