2-Brom-N-methylbenzolsulfonamid

Lieferant: COMBI-BLOCKS INC.

COBBHC-6086-5GEA 493 EUR
COBBHC-6086-5G COBBHC-6086-1G
2-Brom-N-methylbenzolsulfonamid
2-Brom-N-methylbenzolsulfonamid
Formel: C₇H₈BrNO₂S
Molecular Weight: 250,11 g/mol
MDL: MFCD03084756
CAS-Nummer: 98192-14-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR