3-(4-Boc-piperazino)-4-nitroanisol

Lieferant: COMBI-BLOCKS INC.

COBBHC-3091-5GEA 218 EUR
COBBHC-3091-5G COBBHC-3091-1G
3-(4-Boc-piperazino)-4-nitroanisol
3-(4-Boc-piperazino)-4-nitroanisol
Formel: C₁₆H₂₃N₃O₅
MDL: MFCD13195696
CAS-Nummer: 1215205-93-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR