2-Bromisonicotinamid

Lieferant: COMBI-BLOCKS INC.

COBBCA-4201-5GEA 299 EUR
COBBCA-4201-5G COBBCA-4201-25G COBBCA-4201-1G
2-Bromisonicotinamid
2-Bromisonicotinamid
Formel: C₆H₅BrN₂O
MDL: MFCD00234156
CAS-Nummer: 29840-73-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR