1-Ethyl-1H-pyrazol-4-boronsäure

Lieferant: COMBI-BLOCKS INC.

COBBBB-8817-1GEA 187 EUR
COBBBB-8817-1G COBBBB-8817-5G COBBBB-8817-250M
1-Ethyl-1H-pyrazol-4-boronsäure
1-Ethyl-1H-pyrazole-4-boronic acid
Formel: C₅H₉BN₂O₂
MDL: MFCD10703521
CAS-Nummer: 847818-56-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR