4-Mercaptophenylboronsäure

Lieferant: COMBI-BLOCKS INC.

COBBBB-8372-5GEA 385 EUR
COBBBB-8372-5G COBBBB-8372-1G
4-Mercaptophenylboronsäure
4-Mercaptophenylboronsäure
Formel: C₆H₇BO₂S
Molecular Weight: 153,9946 g/mol
MDL: MFCD03093887
CAS-Nummer: 237429-33-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR