N,N-Dimethyl-3-borono-4-methylbenzolsulfonamid

Lieferant: COMBI-BLOCKS INC.

COBBBB-2908-1GEA 377 EUR
COBBBB-2908-1G COBBBB-2908-5G
N,N-Dimethyl-3-borono-4-methylbenzolsulfonamid
N,N-Dimethyl-3-borono-4-methylbenzolsulfonamid
Formel: C₉H₁₄BNO₄S
Molecular Weight: 243,0900 g/mol
MDL: MFCD08056363
CAS-Nummer: 871332-99-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR