4-Ethoxy-3-(trifluormethyl)anilin
Lieferant: COMBI-BLOCKS INC.
COBBAN-1324-5GEA
97.3
EUR
COBBAN-1324-5G
COBBAN-1324-1G
COBBAN-1324-25G
4-Ethoxy-3-(trifluormethyl)anilin
4-Ethoxy-3-(trifluormethyl)anilin
Formel:
C₉H₁₀F₃NO Molecular Weight: 205,18 g/mol |
MDL:
MFCD08056137 CAS-Nummer: 2713-74-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...