Isochinolin-5-sulfonsäure

Lieferant: CHEMPUR

CHMPCC05003.10EA 0 EUR
CHMPCC05003.10 CHMPCC05003.1 CHMPCC05003.50
Isochinolin-5-sulfonsäure
Isochinolin-5-sulfonsäure
Formel: C₉H₇NO₃S
Molecular Weight: 209,22 g/mol
MDL: MFCD00134089
CAS-Nummer: 27655-40-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR