Tubing, PTFE
Lieferant: Bürkle
BURK8804-1012EA 83.79 EUR
BURK8804-1012 BURK8804-0204 BURK8804-0103 BURK8879-0204 BURK8804-0406 BURK8879-0103 BURK8804-0810 BURK8804-0305
Tubing, PTFE
Schläuche PTFE-Schläuche
PTFE, white, translucent.
  • Excellent chemical resistance
  • Very good thermal resistance, autoclavable
  • Free of extractable substances, physiologically neutral, biocompatible
This tubing is characterised by its non stick surface and excellent dielectric properties.
Temperature resistance: −200 to +260 °C