Ball valves for barrels, LaboPlast®
Lieferant: Bürkle
LaboPlast®
227-1108EA 55.4 EUR
227-1108 227-1109 BURK0500-8002 BURK0500-0134
Ball valves for barrels, LaboPlast®
Hähne
PP, black/red
  • Suitable for solidifying media
  • Max. 4 bar pressure at 20 °C
  • Individually adaptable, through various accessories
Ball valves for barrels with thread connections.