Label printers with Brady workstation LAB suite, BMP™ 71
Lieferant: Brady
BRDY306861EA 1129 EUR
BRDY306861 BRDY306864 BRDY306862 BRDY306863
Label printers with Brady workstation LAB suite, BMP™ 71
Drucker Label Printers
The BMP™ 71, one smart printer to print it all.
  • 300 dpi
  • Mono colour
  • Applications include cable and wire identification, electrical identification, facility and safety identification, and laboratory identification
This ultimate Brady Mobile Printer helps you deal with every situation, speeds up your labelling and keeps you going on and on. It prints any die-cut or continuous label - also custom labels - up to 50.8mm in more than 30 different materials. With its full AZERTY keyboard it allows swift typing with both hands and the enlarged full colour screen shows you the whole label at once. Works with a rechargeable Nimh battery or AC power. 2 year Warranty. The BMP™71 comes with the Brady Workstation Product & Wire Identification Software suite, enabling you to immediately start designing professional signs & labels. This suite combines frequently used software apps to create product & cable identification labels.
Materials supported include Freezerbondz Polyester; Heat-shrink Polyolefin Sleeve; Metal Detectable Polyester; Metallised Polyester; Nylon Cloth; Paper; Phosphorescent Polyester; Polyester; Polyester Tag; Polyester with Polyester Overlaminate; Polyethylene Foam Laminate Polyester; Polyethylene Tag; Polyimide; Polypropylene; Polypropylene Tag; Polyvinylfluoride; Reflective Tape; Self-laminating Polyester; Self-laminating Vinyl; StainerBondz Polyester; Static Dissipative Polyester; Tamper-resistant Metallised Vinyl; Tamper-resistant Vinyl; Thermoplastic Polyether Polyurethane; Vinyl; Vinyl Cloth; Vinyl Tag.
Barcode types include Code 39;128 and more through software. Manual cut; Between labels or at end of print job.
Languages: Danish; Dutch; English; Finnish; French; Italian; Norwegian (Bokmål); Polish; Portuguese; Romanian; Spanish; Swedish.
Lieferumfang: This printer kit ships in a hard case with Rechargeable NiMH Battery, AC Adaptor, USB Communications Cable, TLS2200® / HandiMark™ Material Roll Adaptor, Quick Start Guide, Product CD (w/ Printer Driver, Tutorials, Full User's Guide), Cleaning Kit, a Brady Workstation Laboratory Identification Suite, M71C-2000-595-WT, M71-31-423 and M71-R6000.