Label printers with Brady workstation SFID suite, BBP™ 85
Lieferant: Brady
BRDY198571EA 4795 EUR
BRDY198570 BRDY198571 BRDY198566 BRDY198568
Label printers with Brady workstation SFID suite, BBP™ 85
Drucker Label Printers
The BBP™85 Sign and Label Printer allows you to create upto 254 mm labels and signs in multiple colours.
  • Multicolour and monocolour capability
  • Continuous tape and die cut labels
  • Support materials such as phosphorescent polyester; polyester; reflective tape; vinyl
  • Improved interface makes it easy to use
The printer comes with the Brady Workstation Safety and Facility Identification Software suite, enabling you to immediately start designing professional signs and labels. This suite combines frequently used software apps to create safety signs and labels.
Suitable for use in beverage processing, buildings identification, chemical, construction, education, energy utilities, food processing, government, healthcare, manufacturing, maritime, mining, oil and gas, process, pulp & paper, tourism & recreation, warehouse & logistics industries.
Standards: UL; cUL; WEEE RoHS; C-Trick (Australia); GOST R (Russia).
Lieferumfang: This printer includes BBP85 Printer, power cord, USB cable, mouse, ceaning pen, cleaning kit, product CD, driver CD, Ethernet card, quick start guide, Brady workstation safety and facility identification suite.