Pulvertrichter, mit GL-Gewinde oder Normschliff
Lieferant: Bohlender
BOLA
BOHLD738-42EA 142 EUR
BOHLD738-42 BOHLD738-52 BOHLD738-12 BOHLD738-22 BOHLD739-22
Pulvertrichter, mit GL-Gewinde oder Normschliff
Trichter Pulvertrichter
Powder funnels, made of borosilicate glass with outlet pipe of PTFE, connection with GL screw caps of PPS or ground joint.
  • The glass funnel can be fixed in any position
  • Used for reaction vessels with GL threaded connections or grounding sleeves
  • Temperature resistance −20 to +200 °C
  • Universal chemical resistance