3-Amino-3-(4-hydroxyphenyl)propionsäure

Lieferant: BIONET RESEARCH (CHEMPUR)

BIOT8R-0644.500EA 0 EUR
BIOT8R-0644.500 BIOT8R-0644.5 BIOT8R-0644.1
3-Amino-3-(4-hydroxyphenyl)propionsäure
3-Amino-3-(4-hydroxyphenyl)propionsäure
Formel: C₉H₁₁NO₃
MDL: MFCD00181810
CAS-Nummer: 6049-54-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR