Sucrose solutions for refractometer calibration
Lieferant: Bellingham + Stanley
BELLSS20EA 144 EUR
BELLSS20 BELLSS30 BELLSS55 BELLSS00 BELLSS10 BELLSS35 BELLSS12 BELLSS115 BELLSS45 BELLSS15 BELLSS125 BELLSS25 BELLSS05 BELLSS112 BELLSS60 BELLSS075 BELLSS40 BELLSS50
Sucrose solutions for refractometer calibration
Standards Refraktometrie-Standards
Sucrose solutions are an extremely easy to use medium for verifying and calibrating refractometers measuring in the Brix scale, as no special consideration is required when testing instruments at ambient temperature fitted with ATC.
  • Uncertainty: ±0,02 °Brix
  • Traceable to NIST and ICUMSA
  • Supplied in 15 ml plastic dropper bottles for ease of use
  • Sucrose solutions have a limited shelf life of 6 weeks
Sucrose solutions are manufactured on a %weight/weight basis and are calibrated in accordance with EN ISO/IEC 17025:2005.