Pulvertrichter
Lieferant: Bel-Art Products, a part of SP
SCIENCEWARE®
BELAH146600065EA 149 EUR
BELAH146600065 BELAH146600100 BELAH146600150 BELAH146600080
Pulvertrichter
Trichter Pulvertrichter
PP, translucent.
  • Very good chemical resistance
  • Very good temperature resistance
  • Autoclavable
Specially designed for transferring powders. These funnels have 60º angle and a wide stem for clog-free filling.
  • Very good chemical resistance
  • Very good temperature resistance
  • Autoclavable
Specially designed for transferring powders. These funnels have 60º angle and a wide stem for clog-free filling.