Platelet-derived growth factor-BB (PDGF-BB), Corning
Lieferant: Corning
BDAA356051EA 0 EUR
BDAA356051
Platelet-derived growth factor-BB (PDGF-BB), Corning
Zellkultur-Ergänzungen und -Zusätze
Platelet-Derived Growth Factor-BB (PDGF-BB), human recombinant, is a dimeric glycoprotein composed of two A and/or B chains.
Applications include culture of various cell types derived from connective tissue. It can also be used to study chemotaxis, wound healing, and bone repair.
Tested for ability to stimulate proliferation of Swiss albino mouse fibroblasts. Filtered (0,2 µm membrane) and tested and found negative for the presence of bacteria, fungi, and mycoplasma.
It is the principal mitogen inserum for mesenchymal cells.