7S Nerve Growth Factor (NGF)
Lieferant: Corning
Corning®
BDAA354009EA 0 EUR
BDAA354009
7S Nerve Growth Factor (NGF)
Zellkultur, Ergänzungen und Zusätze
Useful for maintenance and differentiation of sympathetic and sensory neurons, and neuronal cells in vitro. NGF also has wound healing activity and can be used in degenerative brain disease and nerve injury models.
Tested for ability to stimulate neurite outgrowth of chick dorsal root ganglia. Filtered (0,2 µm membrane) and tested and found negative for the presence of bacteria, fungi, and mycoplasma.