The IVF dish with four wells and a single lid is a single-use device for manipulation and observation of ova and embryos.
- Performance tested: each lot of Falcon® IVF products is subjected to a performance test for growth with a mammalian cell line
- Embryotoxicity: each lot tested using one-cell mouse embro assay
- Nonpyrogenic: product is less than 0.1 EU/ml or 5 EU/device
- Irradiated to meet a minimum sterility level (SAL) of 10⁻⁶
- Cytoxicity resins to USP class VI
- Dish bottoms raised slightly to reduce abrasions and ensure clarity
- Bevel around 4-well plate interior perimeter to prevent settling in corner
- Lids designed for aseptic manipulation and venting
- Packaged in peel-open, medical-style packaging
The Falcon® IVF 4-Well dish design features a sterile, flat, optically clear, single-use device for optimum manipulation and observation of ova and embryos. The Falcon® IVF 4-Well single lid dish has numbered wells and a large writing patch for clear sample identification. Plates are packaged in individual, peel-open trays for sterile presentation.
Standards: Manufactured in compliance with current version of EN ISO 13485 standard.
FDA Class II medical Device and CE marked in compliance with the European Medical Devices Directive 93/42/EEC as class IIa medical device.
Falcon® IVF products are:
Non-embryotoxic*
Non-pyrogenic
Non-cytotoxic
Non-genotoxic
Non-mutagenic
*Falcon IVF lids are not tested or certified for embryotoxicity.
Verpackung: Packaged in peel-open, medical-style packaging.