2,4-Dinitroanisol ≥98%

Lieferant: Thermo Scientific
Achtung

B21542.22EA 125 EUR
B21542.22 B21542.36 B21542.14
2,4-Dinitroanisol ≥98%
2,4-Dinitroanisol

Purity: 98%
CAS No.: 119-27-7
Molecular Formula: C7H6N2O5
Formula Weight: 198.14
Melting Point: 88-94°
Boiling Point: 207°/12mm
UN No.: 2811
MDL No.: MFCD00035745

Formel: C₇H₆N₂O₅
Molecular Weight: 198,14 g/mol
Siedepunkt: 351 °C (1013 hPa)
Schmelzpunkt: 88 °C
Dichte: 1,34 g/cm³ (20 °C)
MDL: MFCD00035745
CAS-Nummer: 119-27-7
EINECS: 204-310-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR