Pulvertrichter, Azlon®
Lieferant: SciLabware
AZLOFWC206EA 53.5 EUR
AZLOFWC206 AZLOFWC212 AZLOFWC210 221-0278
Pulvertrichter, Azlon®
Trichter Pulvertrichter
Funnels, made of PP, with parallel stem minimises 'bridging' of powder to improve flow.
  • Lightweight and unbreakable
  • Two largest sizes have vertical sided tops
  • Good chemical and temperature resistance