4-Ethoxy-3-(trifluormethyl)anilin 98%

Lieferant: Apollo Scientific

APOSPC302332-5GEA 43.7 EUR
APOSPC302332-5G APOSPC302332-1G APOSPC302332-10G
4-Ethoxy-3-(trifluormethyl)anilin 98%
4-Ethoxy-3-(trifluormethyl)anilin
Formel: C₉H₁₀F₃NO
Molecular Weight: 205,18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08056137
CAS-Nummer: 2713-74-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR