3-Chlor-5-fluor-4-methoxy-2-methylbenzaldehyd 97%

Lieferant: Apollo Scientific

APOSPC302291-1GEA 121 EUR
APOSPC302291-1G APOSPC302291-10G APOSPC302291-5G
3-Chlor-5-fluor-4-methoxy-2-methylbenzaldehyd 97%
3-Chloro-5-fluoro-4-methoxy-2-methylbenzaldehyde
Formel: C₉H₈ClFO₂
Molecular Weight: 202,61 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22201016
CAS-Nummer: 1373920-69-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR