3-Fluor-2-methylphenylboronsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
APOSPC10380-1GEA
51
EUR
APOSPC10380-1G
APOSPC10380-5G
3-Fluor-2-methylphenylboronsäure 98%
3-Fluor-2-methylphenylboronsäure
Formel:
C₇H₈BFO₂ Molecular Weight: 153,95 g/mol Schmelzpunkt: 142…144 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD07368679 CAS-Nummer: 163517-61-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...