4-Hydroxy-2-(trifluormethyl)benzonitril 98%

Lieferant: Apollo Scientific
Achtung

APOSPC10356-1GEA 143 EUR
APOSPC10356-1G APOSPC10356-5G APOSPC10356-250MG
4-Hydroxy-2-(trifluormethyl)benzonitril 98%
4-Hydroxy-2-(trifluormethyl)benzonitril
Formel: C₈H₄F₃NO
Molecular Weight: 187,12 g/mol
Schmelzpunkt: 117…124 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06797340
CAS-Nummer: 320-42-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR