2-Bromisonicotinamid 97%

Lieferant: Apollo Scientific

APOSOR9565-5GEA 259 EUR
APOSOR9565-5G APOSOR9565-25G APOSOR9565-1G
2-Bromisonicotinamid 97%
2-Bromisonicotinamid
Formel: C₆H₅BrN₂O
Schmelzpunkt: 188…191 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00234156
CAS-Nummer: 29840-73-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR