1-Ethyl-1H-pyrazol-4-boronsäure 98%

Lieferant: Apollo Scientific

APOSOR907490-1GEA 183 EUR
APOSOR907490-1G APOSOR907490-5G
1-Ethyl-1H-pyrazol-4-boronsäure 98%
1-Ethyl-1H-pyrazole-4-boronic acid
Formel: C₅H₉BN₂O₂
Molecular Weight: 139,9500 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10703521
CAS-Nummer: 847818-56-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR