4-Amino-1-Boc-piperidin-4-carbonsäure 97%

Lieferant: Apollo Scientific

APOSOR480215-5GEA 94.5 EUR
APOSOR480215-5G APOSOR480215-25G
4-Amino-1-Boc-piperidin-4-carbonsäure 97%
4-Amino-1-Boc-piperidin-4-carbonsäure
Formel: C₁₁H₂₀N₂O₄
Molecular Weight: 244,29 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01318728
CAS-Nummer: 183673-71-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR