N-tert-Butyl-3-boronobenzolsulfonamid 97%

Achtung

APOSOR4028-500MGEA 265 EUR
APOSOR4028-250MG APOSOR4028-500MG APOSOR4028-1G
N-tert-Butyl-3-boronobenzolsulfonamid 97%
N-tert-Butyl-3-boronobenzolsulfonamid
Formel: C₁₀H₁₆BNO₄S
Molecular Weight: 257,1143 g/mol
Schmelzpunkt: 120…124 °C
Lagertemperatur: Kühlschrank
MDL: MFCD06659842
CAS-Nummer: 221290-14-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR