Oxazol-5-methanol 97%

Achtung

APOSOR3790-5GEA 0 EUR
APOSOR3790-5G APOSOR3790-1G APOSOR3790-250MG
Oxazol-5-methanol 97%
Oxazol-5-methanol
Formel: C₄H₅NO₂
Molecular Weight: 99,0880 g/mol
Schmelzpunkt: 27…28 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD09027272
CAS-Nummer: 127232-41-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR