Methyl-4-amino-3-iodbenzoat 95+%

Lieferant: Apollo Scientific

APOSOR346462-25GEA 152 EUR
APOSOR346462-25G APOSOR346462-5G
Methyl-4-amino-3-iodbenzoat 95+%
Methyl-4-amino-3-iodbenzoat
Formel: C₈H₈INO₂
Molecular Weight: 277,0600 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00090423
CAS-Nummer: 19718-49-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR