2-Chlor-2'-methoxyacetanilid 95%

Lieferant: Apollo Scientific

APOSOR33345-1GEA 161 EUR
APOSOR33345-1G APOSOR33345-5G APOSOR33345-25G
2-Chlor-2'-methoxyacetanilid 95%
2-Chlor-2'-methoxyacetanilid
Formel: C₉H₁₀ClNO₂
Molecular Weight: 199,6300 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00095394
CAS-Nummer: 55860-22-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR