4-Mercaptophenylboronsäure

Lieferant: Apollo Scientific

APOSOR315062-5GEA 357 EUR
APOSOR315062-5G APOSOR315062-1G
4-Mercaptophenylboronsäure
4-Mercaptophenylboronsäure
Formel: C₆H₇BO₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD03093887
CAS-Nummer: 237429-33-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR