3-(Chlormethyl)benzoesäure

Lieferant: Apollo Scientific

Gefahr

APOSOR2331-1GEA 14.7 EUR
APOSOR2331-1G APOSOR2331-5G APOSOR2331-25G
3-(Chlormethyl)benzoesäure
3-(Chloromethyl)benzoic acid
Formel: C₈H₇ClO₂
Molecular Weight: 170,59
Schmelzpunkt: 136…138 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00191922
CAS-Nummer: 31719-77-4
UN: 3261
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR