4-(Ethylamino)benzoesäure

Lieferant: Apollo Scientific
Achtung

APOSOR22095-1GEA 23.4 EUR
APOSOR22095-1G APOSOR22095-10G APOSOR22095-5G
4-(Ethylamino)benzoesäure
4-(Ethylamino)benzoic acid
Formel: C₉H₁₁NO₂
Schmelzpunkt: 175…177 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00102118
CAS-Nummer: 7409-09-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR