2-Brom-N-methylbenzolsulfonamid 98%

Lieferant: Apollo Scientific

Achtung

APOSOR11471-5GEA 473 EUR
APOSOR11471-5G APOSOR11471-1G
2-Brom-N-methylbenzolsulfonamid 98%
2-Brom-N-methylbenzolsulfonamid
Formel: C₇H₈BrNO₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD03084756
CAS-Nummer: 98192-14-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR