Inosin
Lieferant: Apollo Scientific
APOSBII1001-100GEA
125
EUR
APOSBII1001-100G
APOSBII1001-10G
APOSBII1001-5G
Inosin
Inosin
Formel:
C₁₀H₁₂N₄O₅ Molecular Weight: 268,23 g/mol Schmelzpunkt: 212…213 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00066770 CAS-Nummer: 58-63-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...