L(-)-Fucose

Lieferant: Apollo Scientific
Achtung

APOSBI3933-25GEA 256 EUR
APOSBI3933-25G APOSBI3933-10G APOSBI3933-100G
L(-)-Fucose
L(-)-Fucose
Formel: C₆H₁₂O₅
Molecular Weight: 164,16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00069812
CAS-Nummer: 2438-80-4
EINECS: 219-452-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR