Brilliant III Ultra-Fast SYBR® green ROX QPCR master mix
Lieferant: Agilent Technologies
Brilliant III
AGLS600892EA 355 EUR
AGLS600892 AGLS600904 AGLS600903 AGLS600889
Brilliant III Ultra-Fast SYBR® green ROX QPCR master mix
Nukleinsäure-Reagenzien qPCR/RT-qPCR Enzyme und Kits
High performance, ultra-sensitive, SYBR® Green QPCR master mix reagent with ROX concentration for reliable quantification across a wide range of targets and templates.
  • Reliable and reproducible data
  • Novel fast Taq mutant for qPCR results in under 35 minutes Resistant to common QPCR inhibitors ( whole blood, humic acid, NaCl)
  • Enhanced rapid hot start capability saves time and reduces primer-dimer formation
  • Convenient pre-blended formulations compatible with any SYBR Green dye
Available with low or high ROX concentrations.