Brilliant III Ultra-fast probe ROX qPCR master mix
Lieferant: Agilent Technologies
AGLS600888EA 312 EUR
AGLS600888 AGLS600899 AGLS600898 AGLS600890
Brilliant III Ultra-fast probe ROX qPCR master mix
Nukleinsäure-Reagenzien qPCR/RT-qPCR Enzymes and Kits
High performance, ultra-sensitive, probe QPCR master mix reagent with high or low ROX concentration for reliable quantification across a wide range of targets and templates.
  • Resistant to common QPCR inhibitors (whole blood, humic acid, NaCl)
  • Extremely fast while maintaining sensitivity
  • Optimized fast cycling formulation ensures reliable and reproducible data with shorter run times
Brilliant III Ultra-Fast maintains amplification efficiency, R2, dynamic range and detection sensitivity all while shortening the total cycling time by ~60% compared to standard cycling.
High performance, ultra-sensitive, probe QPCR master mix reagent with high/low ROX concentration for reliable quantification across a wide range of targets and templates. Convenient pre-blended formulations compatible with any sequence-specific probe detection chemistry.
Bitte beachten For research use only. Not for use in diagnostic procedures.