Brilliant III Probe MM, Agilent Technologies
Lieferant: Agilent Technologies
AGLS600885EA 5571 EUR
AGLS600885 AGLS600884 AGLS600881 732-3798
Brilliant III Probe MM, Agilent Technologies
Nukleinsäure-Reagenzien qPCR/RT-qPCR Enzymes and Kits
Brilliant III QPCR and QRT-PCR master mixes increase overall sensitivity and speed through the novel, quick-activating hot start and the engineered Taq-mutant.
  • Extremely fast while maintaining sensitivity
  • Greater resistance to common QPCR inhibitors (whole blood or NaCl)
  • Optimized fast cycling formulation ensures reliable and reproducible data with shorter run times
  • Convenient pre-blended formulations compatible with any sequence-specific probe detection chemistry
Brilliant III Ultra-Fast maintains amplification efficiency, R2, dynamic range and detection sensitivity all while shortening the total cycling time by approximately 60% compared to standard cycling.
Bitte beachten For research use only. Not for use in diagnostic procedures.