Brilliant II probe MM
Lieferant: Agilent Technologies
AGLS600810EA 799 EUR
AGLS600810 AGLS600809 AGLS600819 AGLS600818 AGLS600827 AGLS600815 AGLS600804
Brilliant II probe MM
Nukleinsäure-Reagenzien qPCR/RT-qPCR Enzymes and Kits
Brilliant II QPCR and QRT-PCR reagents were developed for improved sensitivity of detection, ensuring reproducible quantification even at low target concentrations.
  • Greater flexibility for use with numerous different templates and targets
  • Developed on the Mx QPCR Systems
  • Passive reference dye to maximise performance on different instrument platforms
  • Multiplex kit for improved results on multiple targets
For faster experiments with earlier Ct's and greater reproducibility, Brilliant III Ultra-Fast master mixes can be used.
Bitte beachten For research use only. Not for use in diagnostic procedures.