Brilliant II probe MM with ROX
Lieferant: Agilent Technologies
AGLS600806EA 562 EUR
AGLS600806 AGLS600805
Brilliant II probe MM with ROX
Nukleinsäure-Reagenzien qPCR/RT-qPCR Enzyme und Kits
Brilliant II QPCR and QRT-PCR reagents were developed to maintain the high quality of original Brilliant product line, but with a significant improvement in sensitivity of detection, ensuring reproducible quantification even at low target concentrations.
  • Formulations available with low and high concentrations of ROX reference dye
  • Improved performance with earlier Ct detection across a wide dynamic range
  • Increased sensitivity of detection to low copy numbers
With the addition of ROX reference dye to our master mix, the reaction setup process is streamlined.
Bitte beachten For research use only. Not for use in diagnostic procedures.