PfuUltra high-fidelity DNA polymerase
Lieferant: Agilent Technologies
AGLS600389EA 1220 EUR
AGLS600389 AGLS600630 AGLS600387 AGLS600385 AGLS600396 AGLS600384 AGLS600394 AGLS600382 AGLS600392 AGLS600380 AGLS600390
PfuUltra high-fidelity DNA polymerase
Nukleinsäure-Reagenzien Enzyme und Kits für Endpunkt-PCR
PfuUltra High-Fidelity DNA Polymerase AD features a reformulated buffer system for increased economy with the same high performance as the original PfuUltra DNA Polymerase for robust, ultra-high-fidelity PCR.
PfuUltra Hotstart DNA Polymerase features a genetically engineered mutant of Pfu DNA Polymerase and the ArchaeMaxx Polymerase Enhancing Factor and is formulated for hot start activity.
PfuUltra Hotstart DNA Polymerase AD features a reformulated buffer system and novel hot start mechanism for increased economy with the same high performance as the original PfuUltra Hotstart DNA Polymerase.
PfuUltra Hotstart PCR Master Mix is a 2X formulation of PfuUltra Hotstart DNA Polymerase, an optimsed PCR reaction buffer, magnesium, and dNTPs.