Alien QRT-PCR
Lieferant: Agilent Technologies
AGLS300600EA 466 EUR
AGLS300600 AGLS300602 AGLS300604
Alien QRT-PCR
Nukleinsäure-Reagenzien qPCR/RT-qPCR Enzymes and Kits
Alien QRT-PCR Inhibitor Alert is an external control to detect inhibitors in quantitative RT-PCR RNA.
  • Superior external control to detect inhibitors in RNA samples
  • No notable nucleic acid homology
  • Can be used to detect inhibitors in either 1-Step (single-tube) or 2-Step (two-tube) QRT-PCR assays utilising SYBR Green dye for detection
Alien QRT-PCR Inhibitor Alert is a superior external control to detect inhibitors in RNA samples for quantitative RT-PCR assays. The Alien RNA transcript is an RNA molecule transcribed in vitro with no notable nucleic acid homology. It is used to compare amplification of known quantities of Alien RNA.
The threshold cycle (Ct) for amplification of the Alien RNA is measured in reference reactions holding the Alien RNA transcript alone. It can be used to detect inhibitors in either 1-Step (single-tube) or 2-Step (two-tube) QRT-PCR assays utilising SYBR Green dye for detection.
Bitte beachten For research use only. Not for use in diagnostic procedures.