Retroviral gene expression system, ViraPort
Lieferant: Agilent Technologies
AGLS217569EA 557 EUR
AGLS217569 AGLS217568 AGLS240029 AGLS217567 AGLS217566 AGLS240030 AGLS217561 AGLS217563 AGLS217570
Retroviral gene expression system, ViraPort
Protein- Expression Protein- Expressionsysteme
The ViraPort retroviral gene expression system is a comprehensive line of gene delivery products providing transduction efficiencies approaching 100% in a wide range of mitotic cells.
  • Powerful system for functional cloning
  • Copy number controlled by multiplicity of infection
  • Create high-titer retroviral virions for library screening
Bitte beachten For Research Use Only. Not for Use in Diagnostic Procedures.