Nickelsulfat Hexahydrat 99% zur Analyse
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Nickel(II)sulfat Hexahydrat
ACRO211081000EA
31.5
EUR
ACRO211081000
ACRO211080025
ACRO211080100
ACRO211085000
Nickelsulfat Hexahydrat 99% zur Analyse
Nickelsulfat Hexahydrat
Formel:
NiSO₄·6H₂O Molecular Weight: 262,85 g/mol Schmelzpunkt: 53 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149813 CAS-Nummer: 10101-97-0 EINECS: 232-104-9 UN: 3077 ADR: 9,III Merck Index: 13,06541 |
Spezifikation Testergebnisse
Appearance | Blue-green Crystals |
Titration Complexometric | ≥98.5 % |
pH | 4 to 6 (5 % at 20°C) |
Chloride (Cl) | ≤50 ppm |
Arsenic (As) | ≤10 ppm |
Cadmium (Cd) | ≤50 ppm |
Cobalt (Co) | ≤100 ppm |
Copper (Cu) | ≤20 ppm |
Iron (Fe) | ≤50 ppm |
Lead (Pb) | ≤20 ppm |
Zinc (Zn) | ≤50 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...