Di(2-pyridyl)keton 98%
Lieferant: Thermo Scientific
|
Achtung
|
ACRO155670250EA
388
EUR
ACRO155670250
ACRO155670050
Di(2-pyridyl)keton 98%
Di(2-pyridyl)keton
Formel:
C₁₁H₈N₂O Molecular Weight: 184,20 g/mol Schmelzpunkt: 53…55 °C Flammpunkt: >110 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006288 CAS-Nummer: 19437-26-4 EINECS: 243-067-3 |
Spezifikation Testergebnisse
Appearance | Light yellow to beige to brown Crystalline powder or crystals and/or chunks |
Infrared spectrum | Conforms |
Melting point | 49 to 54 °C |
GC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...