Recombinat Protein,Human DR3 TNFRSF25 Protein, Fc Tag,Human DR3, Fc Tag (TN5-H5258) is expressed from human 293 cells (HEK293). It contains AA Gln 25 - Gln 199 (Accession AAI17190).This protein carries a human IgG1 Fc tag at the C-terminus. The prot 1 * 100 µG
Lieferant: ACROBIOSYSTEMS INC